Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927650 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110918.1 | 4 | 679 | |||
GenBank mRNA | AL110259.1 | - | 4 | 679 |