Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FER SIRNA Assays ›
          Gene Symbol:
          FER
          Gene Name:
          FER tyrosine kinase
          Gene Aliases:
          PPP1R74, TYK3, p94-Fer
          Chromosome Location:
          Chr.5: 108746085 - 109196841 on Build GRCh38
          UniGene ID:
          Hs.107418
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n339107

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 404.00
          Your Price
          Online offer: 391.65
          391.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FER
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001308028.1 NP_001294957.1 18 6605
          NM_001308031.1 NP_001294960.1 11 5555
          NM_005246.3 NP_005237.2 20 6991
          XM_011543276.2 XP_011541578.1 12 5807
          GenBank mRNA AK123847.1 - 1 220

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          mitotic cell cycle
          regulation of protein phosphorylation
          protein phosphorylation
          chemotaxis
          cell adhesion
          cell proliferation
          positive regulation of cell proliferation
          regulation of lamellipodium assembly
          regulation of fibroblast migration
          peptidyl-tyrosine phosphorylation
          cytokine-mediated signaling pathway
          cell differentiation
          positive regulation of cell migration
          positive regulation of actin filament polymerization
          actin cytoskeleton reorganization
          response to lipopolysaccharide
          cellular response to insulin stimulus
          negative regulation of mast cell activation involved in immune response
          substrate adhesion-dependent cell spreading
          cellular response to reactive oxygen species
          extracellular matrix-cell signaling
          intracellular signal transduction
          cellular response to macrophage colony-stimulating factor stimulus
          response to platelet-derived growth factor
          insulin receptor signaling pathway via phosphatidylinositol 3-kinase
          peptidyl-tyrosine autophosphorylation
          Fc-epsilon receptor signaling pathway
          Kit signaling pathway
          regulation of epidermal growth factor receptor signaling pathway
          tyrosine phosphorylation of Stat3 protein
          regulation of mast cell degranulation
          cell-cell adhesion mediated by cadherin
          innate immune response
          protein autophosphorylation
          platelet-derived growth factor receptor signaling pathway
          diapedesis
          positive regulation of NF-kappaB transcription factor activity
          interleukin-6-mediated signaling pathway
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          epidermal growth factor receptor binding
          protein binding
          ATP binding
          protein phosphatase 1 binding
          lipid binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0