Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017028105.1 | XP_016883594.1 | 19 | 7411 | ||
XM_017028107.1 | XP_016883596.1 | 12 | 7501 | |||
GenBank mRNA | AK022738.1 | - | 1 | 2196 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CN360205.1 | - | 1 | 349 | Chr. 20 - 5998405 - 5999025 [+] on Build GRCh38 |