Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
Unity Lab Services
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LOC101927583 SIRNA Assays ›
Gene Symbol:
LOC101927583
Gene Name:
uncharacterized LOC101927583
Gene Aliases:
-
Chromosome Location:
Chr.12: 57618405 - 57621903 on Build GRCh38
UniGene ID:
Hs.624788
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
n344051
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LOC101927583
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_120450.1
1
1230
GenBank mRNA
BC050035.1
-
1
1230
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0