Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HFE SIRNA Assays ›
Gene Symbol:
HFE
Gene Name:
hemochromatosis
Gene Aliases:
HFE1, HH, HLA-H, MVCD7, TFQTL2
Chromosome Location:
Chr.6: 26087281 - 26096216 on Build GRCh38
UniGene ID:
Hs.233325
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
n349309
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
404.00
Your Price
Online offer:
391.65
391.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HFE
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_011514543.2
XP_011512845.1
7
2437
GenBank mRNA
AF147392.1
-
1
143
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
Transcript Location
GenBankmRNA
DA957218.1
-
1
171
Chr. 6 - 26095509 - 26096070 [+] on Build GRCh38
GenBankmRNA
T59431.1
-
73
Chr. - - [-] on Build
GenBankmRNA
BX107211.1
-
1
242
Chr. 6 - 26095471 - 26096083 [+] on Build GRCh38
Back To Top
More Information
Panther Classification:
Molecular Function -
major histocompatibility complex protein
Gene Ontology Categories:
Function(s)
Process(es)
antigen processing and presentation of peptide antigen via MHC class I
negative regulation of T cell antigen processing and presentation
negative regulation of T cell cytokine production
protein complex assembly
cellular iron ion homeostasis
acute-phase response
immune response
female pregnancy
response to iron ion
cellular response to iron ion starvation
positive regulation of gene expression
positive regulation of pathway-restricted SMAD protein phosphorylation
antigen processing and presentation
BMP signaling pathway
positive regulation of protein binding
negative regulation of proteasomal ubiquitin-dependent protein catabolic process
hormone biosynthetic process
positive regulation of receptor-mediated endocytosis
iron ion homeostasis
multicellular organismal iron ion homeostasis
cellular response to iron ion
positive regulation of peptide hormone secretion
liver regeneration
iron ion import into cell
negative regulation of receptor binding
positive regulation of receptor binding
positive regulation of ferrous iron import into cell
negative regulation of antigen processing and presentation of endogenous peptide antigen via MHC class I
positive regulation of ferrous iron binding
positive regulation of transferrin receptor binding
response to iron ion starvation
regulation of protein localization to cell surface
negative regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
negative regulation of receptor activity
positive regulation of receptor activity
negative regulation of CD8-positive, alpha-beta T cell activation
antigen binding
receptor binding
protein binding
beta-2-microglobulin binding
co-receptor binding
peptide antigen binding
transferrin receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-sg7j2:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0