Search
Search

LOC105375531
TBXAS1| Gene Symbol: LOC105375531 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK000794.1 | - | 1 | 1877 | ||
| Gene Symbol: TBXAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017012570.1 | XP_016868059.1 | 11 | 5050 | ||