Search
Search

FRY| Gene Symbol: FRY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006719749.3 | XP_006719812.1 | 63 | 13899 | ||
| XM_017020303.1 | XP_016875792.1 | 63 | 14376 | |||
| XM_017020304.1 | XP_016875793.1 | 62 | 12800 | |||
| XM_017020305.1 | XP_016875794.1 | 63 | 14395 | |||
| XM_017020306.1 | XP_016875795.1 | 42 | 9537 | |||
| XM_017020307.1 | XP_016875796.1 | 36 | 8890 | |||
| GenBank mRNA | BC036296.1 | - | 1 | 457 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG743355.1 | - | 1 | 456 | Chr. 13 - 32298257 - 32299084 [+] on Build GRCh38 | |