Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SIRT2 SIRNA Assays ›
Gene Symbol:
SIRT2
Gene Name:
sirtuin 2
Gene Aliases:
SIR2, SIR2L, SIR2L2
Chromosome Location:
Chr.19: 38878555 - 38899862 on Build GRCh38
UniGene ID:
Hs.466693
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
n359695
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SIRT2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_006723111.1
XP_006723174.1
2
237
GenBank mRNA
AK094810.1
-
1
838
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
chromatin silencing at rDNA
chromatin silencing
chromatin silencing at telomere
transcription, DNA-templated
protein ADP-ribosylation
protein deacetylation
autophagy
mitotic nuclear division
regulation of exit from mitosis
negative regulation of cell proliferation
negative regulation of autophagy
negative regulation of peptidyl-threonine phosphorylation
phosphatidylinositol 3-kinase signaling
gene silencing
histone deacetylation
substantia nigra development
myelination in peripheral nervous system
regulation of myelination
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
cellular response to oxidative stress
peptidyl-lysine deacetylation
cellular response to hepatocyte growth factor stimulus
negative regulation of protein catabolic process
regulation of phosphorylation
negative regulation of apoptotic process
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of DNA binding
protein kinase B signaling
cellular lipid catabolic process
innate immune response
negative regulation of fat cell differentiation
positive regulation of meiotic nuclear division
negative regulation of striated muscle tissue development
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
hepatocyte growth factor receptor signaling pathway
negative regulation of oligodendrocyte differentiation
cell division
meiotic cell cycle
regulation of cell cycle
response to redox state
positive regulation of cell division
positive regulation of attachment of spindle microtubules to kinetochore
negative regulation of transcription from RNA polymerase II promoter in response to hypoxia
cellular response to caloric restriction
negative regulation of oligodendrocyte progenitor proliferation
histone H3 deacetylation
histone H4 deacetylation
cellular response to molecule of bacterial origin
cellular response to hypoxia
cellular response to epinephrine stimulus
tubulin deacetylation
positive regulation of execution phase of apoptosis
positive regulation of oocyte maturation
negative regulation of NLRP3 inflammasome complex assembly
negative regulation of defense response to bacterium
ripoptosome assembly involved in necroptotic process
negative regulation of reactive oxygen species metabolic process
positive regulation of proteasomal ubiquitin-dependent protein catabolic process involved in cellular response to hypoxia
chromatin binding
NAD+ ADP-ribosyltransferase activity
histone deacetylase activity
protein binding
transcription factor binding
zinc ion binding
NAD-dependent histone deacetylase activity
protein deacetylase activity
NAD-dependent protein deacetylase activity
histone acetyltransferase binding
histone deacetylase binding
tubulin deacetylase activity
ubiquitin binding
NAD-dependent histone deacetylase activity (H4-K16 specific)
beta-tubulin binding
NAD+ binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-jg9xr:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline