Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ARRB1 SIRNA Assays ›
          Gene Symbol:
          ARRB1
          Gene Name:
          arrestin beta 1
          Gene Aliases:
          ARB1, ARR1
          Chromosome Location:
          Chr.11: 75260122 - 75351831 on Build GRCh38
          UniGene ID:
          Hs.503284
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n361290

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ARRB1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_004041.4 NP_004032.2 16 6701
          NM_020251.3 NP_064647.1 15 6677
          XM_011545034.2 XP_011543336.1 17 6585
          XM_011545035.2 XP_011543337.1 16 6561
          XM_017017750.1 XP_016873239.1 18 6657
          XM_017017751.1 XP_016873240.1 17 6633
          XM_017017752.1 XP_016873241.1 17 6613
          XM_017017753.1 XP_016873242.1 17 6677
          XM_017017754.1 XP_016873243.1 16 6589
          GenBank mRNA AL157484.1 - 1 1522

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          positive regulation of protein phosphorylation
          G-protein coupled receptor internalization
          positive regulation of receptor internalization
          transcription from RNA polymerase II promoter
          apoptotic process
          phototransduction
          protein transport
          protein ubiquitination
          platelet activation
          negative regulation of protein ubiquitination
          positive regulation of protein ubiquitination
          negative regulation of NF-kappaB transcription factor activity
          positive regulation of protein binding
          negative regulation of interleukin-6 production
          negative regulation of interleukin-8 production
          positive regulation of peptidyl-serine phosphorylation
          negative regulation of GTPase activity
          positive regulation of smooth muscle cell apoptotic process
          positive regulation of Rho protein signal transduction
          positive regulation of histone acetylation
          positive regulation of insulin secretion involved in cellular response to glucose stimulus
          response to antidepressant
          follicle-stimulating hormone signaling pathway
          stress fiber assembly
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          proteasome-mediated ubiquitin-dependent protein catabolic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of GTPase activity
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of ERK1 and ERK2 cascade
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of histone H4 acetylation
          histone acetyltransferase activity
          enzyme inhibitor activity
          GTPase activator activity
          insulin-like growth factor receptor binding
          protein binding
          transcription factor binding
          estrogen receptor binding
          mitogen-activated protein kinase kinase binding
          ubiquitin protein ligase binding
          alpha-1A adrenergic receptor binding
          alpha-1B adrenergic receptor binding
          angiotensin receptor binding
          follicle-stimulating hormone receptor binding
          V2 vasopressin receptor binding
          AP-2 adaptor complex binding
          clathrin adaptor activity
          cysteine-type endopeptidase inhibitor activity involved in apoptotic process
          transcription regulatory region DNA binding
          ion channel binding
          protein phosphorylated amino acid binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0