Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NRG1 SIRNA Assays ›
          Gene Symbol:
          NRG1
          Gene Name:
          neuregulin 1
          Gene Aliases:
          ARIA, GGF, GGF2, HGL, HRG, HRG1, HRGA, MST131, MSTP131, NDF, NRG1-IT2, SMDF
          Chromosome Location:
          Chr.8: 31639222 - 32771716 on Build GRCh38
          UniGene ID:
          Hs.453951
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n364081

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 404.00
          Your Price
          Online offer: 391.65
          391.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NRG1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001159995.2 NP_001153467.1 12 4498
          NM_001159996.2 NP_001153468.1 9 4354
          NM_001159999.2 NP_001153471.1 13 4549
          NM_001160001.2 NP_001153473.1 11 4447
          NM_001160004.2 NP_001153476.1 13 5074
          NM_001322197.1 NP_001309126.1 8 4203
          NM_001322201.1 NP_001309130.1 10 4415
          NM_001322202.1 NP_001309131.1 9 4364
          NM_001322205.1 NP_001309134.1 9 5590
          NM_001322206.1 NP_001309135.1 10 5732
          NM_013956.4 NP_039250.2 13 4956
          NM_013957.4 NP_039251.2 12 4932
          NM_013960.4 NP_039254.1 13 5083
          NM_013964.4 NP_039258.1 12 4941
          XM_005273486.3 XP_005273543.1 8 5575
          XM_005273487.3 XP_005273544.1 8 5566
          XM_006716335.3 XP_006716398.1 9 3941
          XM_011544512.2 XP_011542814.1 13 4739
          XM_017013365.1 XP_016868854.1 12 4653
          XM_017013366.1 XP_016868855.1 12 4644
          XM_017013367.1 XP_016868856.1 11 4449
          XM_017013368.1 XP_016868857.1 11 4786
          XM_017013369.1 XP_016868858.1 9 5558
          XM_017013370.1 XP_016868859.1 8 5538
          GenBank mRNA AF176921.1 - 1 779
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BF217878.1 - 1 156 Chr. 8 - 32766749 - 32767452 [+] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          activation of MAPK activity
          cell morphogenesis
          startle response
          cardiac conduction system development
          ventricular trabecula myocardium morphogenesis
          cell communication
          activation of transmembrane receptor protein tyrosine kinase activity
          nervous system development
          synapse assembly
          peripheral nervous system development
          locomotory behavior
          regulation of blood pressure
          positive regulation of cell proliferation
          embryo development
          neural crest cell development
          regulation of phosphatidylinositol 3-kinase signaling
          positive regulation of phosphatidylinositol 3-kinase signaling
          glucose transport
          peptidyl-tyrosine phosphorylation
          glial cell fate commitment
          chemorepulsion involved in interneuron migration from the subpallium to the cortex
          mammary gland development
          positive regulation of myelination
          activation of protein kinase B activity
          intracellular signal transduction
          ERBB signaling pathway
          ERBB2 signaling pathway
          wound healing
          negative regulation of protein catabolic process
          myelination
          regulation of protein homodimerization activity
          regulation of protein heterodimerization activity
          positive regulation of GTPase activity
          neurotransmitter receptor metabolic process
          positive regulation of axon extension
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of Ras protein signal transduction
          phosphatidylinositol phosphorylation
          phosphatidylinositol-mediated signaling
          neuron fate commitment
          oligodendrocyte differentiation
          positive regulation of peptidyl-tyrosine phosphorylation
          negative regulation of secretion
          positive regulation of striated muscle cell differentiation
          positive regulation of protein kinase B signaling
          cardiac muscle cell differentiation
          cardiac muscle cell myoblast differentiation
          endocardial cell differentiation
          positive regulation of dendritic spine development
          positive regulation of calcineurin-NFAT signaling cascade
          positive regulation of protein targeting to mitochondrion
          positive regulation of protein localization to cell surface
          regulation of cell motility
          positive regulation of cardiac muscle cell differentiation
          negative regulation of neuron migration
          transcription cofactor activity
          protein tyrosine kinase activity
          Ras guanyl-nucleotide exchange factor activity
          cytokine activity
          ErbB-2 class receptor binding
          integrin binding
          growth factor activity
          transmembrane receptor protein tyrosine kinase activator activity
          receptor tyrosine kinase binding
          ErbB-3 class receptor binding
          chemorepellent activity
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0