Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318047.1 | NP_001304976.1 | 23 | 3471 | ||
GenBank mRNA | AF029325.1 | - | 1 | 239 | ||
AK300385.1 | - | 23 | 3472 |