Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199469.1 | NP_001186398.1 | 12 | 2035 | ||
NM_020701.3 | NP_065752.1 | 11 | 1969 | |||
GenBank mRNA | AK024035.1 | - | 1 | 151 |