Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MELTF SIRNA Assays ›
          Gene Symbol:
          MELTF
          Gene Name:
          melanotransferrin
          Gene Aliases:
          CD228, MAP97, MFI2, MTF1, MTf
          Chromosome Location:
          Chr.3: 197001740 - 197029816 on Build GRCh38
          UniGene ID:
          Hs.184727
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n373031

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MELTF
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005929.5 NP_005920.2 16 3707
          XM_006713643.3 XP_006713706.2 16 3784
          XM_011512850.2 XP_011511152.1 16 3787
          GenBank mRNA BC034933.1 - 1 681
          BC068085.1 - 1 453
          BC082761.1 - 1 453
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BM757703.1 - 1 331 Chr. 3 - 197001783 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM758731.1 - 1 331 Chr. 3 - 197001793 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM756635.1 - 1 331 Chr. 3 - 197001782 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM758279.1 - 1 331 Chr. 3 - 197001787 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM758746.1 - 1 331 Chr. 3 - 197001783 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM757665.1 - 1 331 Chr. 3 - 197001774 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM758491.1 - 1 331 Chr. 3 - 197001772 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM750636.1 - 1 331 Chr. 3 - 197001769 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM757689.1 - 1 331 Chr. 3 - 197001773 - 197002326 [-] on Build GRCh38
          GenBankmRNA AA291458.1 - 1 51 Chr. 3 - 197001746 - 197002042 [-] on Build GRCh38
          GenBankmRNA BM758998.1 - 1 331 Chr. 3 - 197001783 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM749707.1 - 1 331 Chr. 3 - 197001792 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM757642.1 - 1 331 Chr. 3 - 197001773 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM757674.1 - 1 331 Chr. 3 - 197001782 - 197002326 [-] on Build GRCh38
          GenBankmRNA BP391592.1 - 1 148 Chr. 3 - 197001741 - 197002142 [-] on Build GRCh38
          GenBankmRNA BM748562.1 - 1 331 Chr. 3 - 197001773 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM743332.1 - 1 331 Chr. 3 - 197001787 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM748658.1 - 1 331 Chr. 3 - 197001771 - 197002326 [-] on Build GRCh38
          GenBankmRNA BU172111.1 - 1 451 Chr. 3 - 197001742 - 197002446 [-] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          positive regulation of plasminogen activation
          regulation of cell proliferation
          iron ion homeostasis
          positive regulation of extracellular matrix disassembly
          iron ion import
          negative regulation of substrate adhesion-dependent cell spreading
          iron ion binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline