Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MELTF SIRNA Assays ›
          Gene Symbol:
          MELTF
          Gene Name:
          melanotransferrin
          Gene Aliases:
          CD228, MAP97, MFI2, MTF1, MTf
          Chromosome Location:
          Chr.3: 197001740 - 197029816 on Build GRCh38
          UniGene ID:
          Hs.184727
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n373034

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MELTF
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005929.5 NP_005920.2 16 3837
          XM_006713643.3 XP_006713706.2 16 3914
          XM_011512850.2 XP_011511152.1 16 3917
          GenBank mRNA BC034933.1 - 1 811
          BC068085.1 - 1 583
          BC082761.1 - 1 583
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BM757703.1 - 1 461 Chr. 3 - 197001783 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM758731.1 - 1 461 Chr. 3 - 197001793 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM758746.1 - 1 461 Chr. 3 - 197001783 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM756635.1 - 1 461 Chr. 3 - 197001782 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM757665.1 - 1 461 Chr. 3 - 197001774 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM758491.1 - 1 461 Chr. 3 - 197001772 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM750636.1 - 1 461 Chr. 3 - 197001769 - 197002326 [-] on Build GRCh38
          GenBankmRNA AA291458.1 - 1 181 Chr. 3 - 197001746 - 197002042 [-] on Build GRCh38
          GenBankmRNA BM757689.1 - 1 461 Chr. 3 - 197001773 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM758998.1 - 1 460 Chr. 3 - 197001783 - 197002326 [-] on Build GRCh38
          GenBankmRNA BG769667.1 - 1 704 Chr. 3 - 197001773 - 197002573 [-] on Build GRCh38
          GenBankmRNA BM749707.1 - 1 461 Chr. 3 - 197001792 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM757674.1 - 1 461 Chr. 3 - 197001782 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM757642.1 - 1 461 Chr. 3 - 197001773 - 197002326 [-] on Build GRCh38
          GenBankmRNA BP391592.1 - 1 278 Chr. 3 - 197001741 - 197002142 [-] on Build GRCh38
          GenBankmRNA BM748562.1 - 1 461 Chr. 3 - 197001773 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM743332.1 - 1 461 Chr. 3 - 197001787 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM748658.1 - 1 461 Chr. 3 - 197001771 - 197002326 [-] on Build GRCh38
          GenBankmRNA BU172111.1 - 1 581 Chr. 3 - 197001742 - 197002446 [-] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          positive regulation of plasminogen activation
          regulation of cell proliferation
          iron ion homeostasis
          positive regulation of extracellular matrix disassembly
          iron ion import
          negative regulation of substrate adhesion-dependent cell spreading
          iron ion binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline