siRNA Details
Gene Symbol: LINC01209 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110819.1 | 2 | 1854 | |||
GenBank mRNA | BC041880.1 | - | 2 | 1855 |

Gene Symbol: LINC01209 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110819.1 | 2 | 1854 | |||
GenBank mRNA | BC041880.1 | - | 2 | 1855 |