Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H60c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB284506.1 | - | 6 | 998 | ||
AK014573.1 | - | 3 | 586 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714012.1 | - | 3 | 588 | Chr. 10 - 3257972 - 3267771 [-] on Build GRCm38 |