Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KCNJ9 SIRNA Assays ›
          Gene Symbol:
          Kcnj9
          Gene Name:
          potassium inwardly-rectifying channel, subfamily J, member 9
          Gene Aliases:
          1700085N21Rik, Girk3, Kir3.3, mbGIRK3
          Chromosome Location:
          Chr.1: 172320501 - 172329331 on Build GRCm38
          UniGene ID:
          Mm.261168
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n394216

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Kcnj9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK007011.1 - 1 942
          BC038600.1 - 1 1526
          BC048090.1 - 1 915
          BC069221.1 - 1 450
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA AA198887.1 - 1 449 Chr. 1 - 172320541 - 172321051 [-] on Build GRCm38
          GenBankmRNA BY625078.1 - 1 311 Chr. 1 - 172320506 - 172320911 [-] on Build GRCm38
          GenBankmRNA BY599950.1 - 1 304 Chr. 1 - 172320506 - 172320903 [-] on Build GRCm38
          GenBankmRNA BY606446.1 - 1 290 Chr. 1 - 172320510 - 172320891 [-] on Build GRCm38
          GenBankmRNA BY647506.1 - 1 299 Chr. 1 - 172320506 - 172320900 [-] on Build GRCm38
          GenBankmRNA BY629991.1 - 1 295 Chr. 1 - 172320518 - 172320896 [-] on Build GRCm38
          GenBankmRNA BY642114.1 - 1 306 Chr. 1 - 172320518 - 172320907 [-] on Build GRCm38
          GenBankmRNA BY657857.1 - 1 299 Chr. 1 - 172320502 - 172320900 [-] on Build GRCm38
          GenBankmRNA BY625885.1 - 1 298 Chr. 1 - 172320510 - 172320898 [-] on Build GRCm38
          GenBankmRNA BX517677.1 - 1 450 Chr. 1 - 172320510 - 172321050 [-] on Build GRCm38
          GenBankmRNA BY638130.1 - 1 300 Chr. 1 - 172320506 - 172320901 [-] on Build GRCm38
          GenBankmRNA BY606827.1 - 1 291 Chr. 1 - 172320506 - 172320892 [-] on Build GRCm38
          GenBankmRNA BY659100.1 - 1 315 Chr. 1 - 172320506 - 172320915 [-] on Build GRCm38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-5gpzd:80/100.66.76.172:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline