Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm17252 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033745.1 | - | 2 | 465 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB625443.1 | - | 2 | 465 | Chr. 9 - 35686435 - 35687390 [-] on Build GRCm38 |