Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sox5os3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040519.1 | 1 | 39 | |||
GenBank mRNA | AK006194.1 | - | 1 | 40 | ||
AK016914.1 | - | 1 | 25 |