siRNA Details
Gene Symbol: Gm2087 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102437.1 | 2 | 115 | |||
GenBank mRNA | AK076672.1 | - | 2 | 117 | ||
AV260139.2 | - | 2 | 132 | |||
BB614956.1 | - | 2 | 117 |

Gene Symbol: Gm2087 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102437.1 | 2 | 115 | |||
GenBank mRNA | AK076672.1 | - | 2 | 117 | ||
AV260139.2 | - | 2 | 132 | |||
BB614956.1 | - | 2 | 117 |