Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gatsl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033062.1 | - | 878 | |||
AK035431.1 | - | 894 | ||||
BC026208.1 | - | 279 |