Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CMSS1 SIRNA Assays ›
          Gene Symbol:
          Cmss1
          Gene Name:
          cms small ribosomal subunit 1
          Gene Aliases:
          1110001A06Rik, 2610528E23Rik, 4930572F24Rik, AV099812, AV122629
          Chromosome Location:
          Chr.16: 57302000 - 57606867 on Build GRCm38
          UniGene ID:
          Mm.425976
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n440325

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Cmss1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK003195.1 - 2 220
          AK080386.1 - 2 212
          AK136657.1 - 2 244
          BC038682.1 - 2 135
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BY102771.1 - 2 180 Chr. 16 - 57561564 - 57606803 [-] on Build GRCm38
          GenBankmRNA AI120191.1 - 2 141 Chr. 16 - 57561431 - 57606759 [-] on Build GRCm38
          GenBankmRNA BB682144.1 - 2 213 Chr. 16 - 57561441 - 57606837 [-] on Build GRCm38
          GenBankmRNA BY448449.1 - 2 138 Chr. 16 - 57561436 - 57606762 [-] on Build GRCm38
          GenBankmRNA BB270728.1 - 1 42 Chr. 16 - 57561436 - 57561763 [-] on Build GRCm38
          GenBankmRNA BY721034.1 - 2 240 Chr. 16 - 57561392 - 57606864 [-] on Build GRCm38
          GenBankmRNA CB056860.1 - 2 205 Chr. 16 - 57561408 - 57606821 [-] on Build GRCm38
          GenBankmRNA BY102783.1 - 2 180 Chr. 16 - 57561563 - 57606803 [-] on Build GRCm38
          GenBankmRNA BY105473.1 - 2 223 Chr. 16 - 57561557 - 57606846 [-] on Build GRCm38
          GenBankmRNA BB639315.1 - 2 212 Chr. 16 - 57561475 - 57606836 [-] on Build GRCm38
          GenBankmRNA CA465091.1 - 2 184 Chr. 16 - 57561381 - 57606806 [-] on Build GRCm38
          GenBankmRNA BB272972.1 - 1 23 Chr. 16 - 57561435 - 57561744 [-] on Build GRCm38
          GenBankmRNA BU924933.1 - 2 222 Chr. 16 - 57561431 - 57606846 [-] on Build GRCm38
          GenBankmRNA CA493059.1 - 248 Chr. - - [-] on Build
          GenBankmRNA BF466153.1 - 1 15 Chr. 16 - 57561527 - 57561736 [-] on Build GRCm38
          GenBankmRNA AA549307.1 - 2 157 Chr. 16 - 57561445 - 57606779 [-] on Build GRCm38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0