Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CMSS1 SIRNA Assays ›
          Gene Symbol:
          Cmss1
          Gene Name:
          cms small ribosomal subunit 1
          Gene Aliases:
          1110001A06Rik, 2610528E23Rik, 4930572F24Rik, AV099812, AV122629
          Chromosome Location:
          Chr.16: 57302000 - 57606867 on Build GRCm38
          UniGene ID:
          Mm.425976
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n440326

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Cmss1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK003195.1 - 2 369
          AK080386.1 - 2 361
          AK136657.1 - 2 393
          BC038682.1 - 2 284
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BB682144.1 - 2 362 Chr. 16 - 57561441 - 57606837 [-] on Build GRCm38
          GenBankmRNA AI120191.1 - 2 290 Chr. 16 - 57561431 - 57606759 [-] on Build GRCm38
          GenBankmRNA BY448449.1 - 2 287 Chr. 16 - 57561436 - 57606762 [-] on Build GRCm38
          GenBankmRNA BB270728.1 - 1 191 Chr. 16 - 57561436 - 57561763 [-] on Build GRCm38
          GenBankmRNA BY721034.1 - 2 389 Chr. 16 - 57561392 - 57606864 [-] on Build GRCm38
          GenBankmRNA CB056860.1 - 2 354 Chr. 16 - 57561408 - 57606821 [-] on Build GRCm38
          GenBankmRNA BY102783.1 - 2 329 Chr. 16 - 57561563 - 57606803 [-] on Build GRCm38
          GenBankmRNA BY105473.1 - 2 372 Chr. 16 - 57561557 - 57606846 [-] on Build GRCm38
          GenBankmRNA CA465091.1 - 2 333 Chr. 16 - 57561381 - 57606806 [-] on Build GRCm38
          GenBankmRNA BB639315.1 - 2 361 Chr. 16 - 57561475 - 57606836 [-] on Build GRCm38
          GenBankmRNA BF466153.1 - 1 164 Chr. 16 - 57561527 - 57561736 [-] on Build GRCm38
          GenBankmRNA BY081066.1 - 2 329 Chr. 16 - 57561472 - 57606803 [-] on Build GRCm38
          GenBankmRNA AA549307.1 - 2 306 Chr. 16 - 57561445 - 57606779 [-] on Build GRCm38
          GenBankmRNA BB261551.1 - 1 149 Chr. 16 - 57561435 - 57561721 [-] on Build GRCm38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8kj9j:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0