Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfpq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017658.1 | - | 1 | 328 | ||
AK141302.1 | - | 1 | 1622 | |||
BC033603.1 | - | 1 | 3020 |