Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CSPRS SIRNA Assays ›
          Gene Symbol:
          Csprs Dapk2 Igh-V3609N Igh-V7183 Igh-VJ558 Igh-VS107 Ighg Ighg3 Ighm Ighv1-53 Ighv1-54 Ighv1-61 Ighv1-62 Ighv1-72 Ighv6-6 Ighv7-3 Igk-V1 LOC102642252 LOC105243590 LOC238440 LOC382693 Rac1 Tnpo3 V165-D-J-C mu
          Gene Name:
          component of Sp100-rs
          death-associated protein kinase 2
          immunoglobulin heavy chain (V3609N non-productive)
          immunoglobulin heavy chain (V7183 family)
          immunoglobulin heavy chain (J558 family)
          immunoglobulin heavy chain (S107 family)
          Immunoglobulin heavy chain (gamma polypeptide)
          Immunoglobulin heavy constant gamma 3
          immunoglobulin heavy constant mu
          immunoglobulin heavy variable 1-53
          immunoglobulin heavy variable V1-54
          immunoglobulin heavy variable 1-61
          immunoglobulin heavy variable V1-62
          immunoglobulin heavy variable 1-72
          immunoglobulin heavy variable 6-6
          immunoglobulin heavy variable 7-3
          immunoglobulin kappa chain variable 1 (V1)
          uncharacterized LOC102642252
          Ig heavy chain Mem5-like
          Ig heavy chain V region
          immunoglobulin heavy chain-like
          RAS-related C3 botulinum substrate 1
          transportin 3
          IgM variable region
          Gene Aliases:
          D1Lub1, HSR
          -
          -
          B9-scFv, IgG, IgH, IgVH1(VSG), VH7183, VI24H
          AI893585
          IgG, IgH, VhNZA6
          AU044919, IGHG2A
          IgG3
          AI326478, Igh-6, Igh-M, Igh6, Igm, muH
          AB069917, Igmv23, Jh2, Jh4
          EG211331, Gm4770, VH
          Gm16900, Vh3
          IgVH
          Gm16709, Ighv186-1
          Gm16695, VH10G1
          Gm16842, Igh-VS107, V(11)-djn
          -
          IgH, IgVH
          IgH, Igg1
          -
          -
          AL023026, D5Ertd559e
          5730544L10Rik, C430013M08Rik, C81142, D6Ertd313e, Trn-SR, mKIAA4133
          -
          Chromosome Location:
          Chr.1|NT_162750: 111572 - 163011 on Build GRCm38
          Chromosome Location:
          Chr.9: 66158226 - 66272242 on Build GRCm38
          Chromosome Location:
          -
          Chromosome Location:
          -
          Chromosome Location:
          -
          Chromosome Location:
          -
          Chromosome Location:
          -
          Chromosome Location:
          Chr.12: 113357442 - 113361232 on Build GRCm38
          Chromosome Location:
          Chr.12: 113418826 - 113422730 on Build GRCm38
          Chromosome Location:
          Chr.12: 115158445 - 115158835 on Build GRCm38
          Chromosome Location:
          Chr.12: 115193675 - 115193968 on Build GRCm38
          Chromosome Location:
          Chr.12: 115359140 - 115359433 on Build GRCm38
          Chromosome Location:
          Chr.12: 115371986 - 115372278 on Build GRCm38
          Chromosome Location:
          Chr.12: 115757984 - 115758277 on Build GRCm38
          Chromosome Location:
          Chr.12: 114434788 - 114435087 on Build GRCm38
          Chromosome Location:
          Chr.12: 114153180 - 114153644 on Build GRCm38
          Chromosome Location:
          -
          Chromosome Location:
          -
          Chromosome Location:
          -
          Chromosome Location:
          -
          Chromosome Location:
          -
          Chromosome Location:
          Chr.5: 143505481 - 143528116 on Build GRCm38
          Chromosome Location:
          Chr.6: 29540827 - 29610110 on Build GRCm38
          Chromosome Location:
          -
          UniGene ID:
          Mm.362644
          Mm.304472
          Mm.342177
          Mm.342177
          Mm.246497
          Mm.359198
          Mm.342177
          Mm.342177
          Mm.304472
          Mm.335723
          Mm.313452
          Mm.390518
          Mm.313465
          Mm.304472
          Mm.422809
          Mm.359198
          Mm.304143
          Mm.351752
          Mm.390491
          Mm.342177
          Mm.313447
          Mm.292510
          Mm.276381
          Mm.335723
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID n443632

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Csprs
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA L10423.1 - 347
          Gene Symbol: Dapk2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AB058015.1 - 344
          Gene Symbol: Igh-V3609N
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA Z37502.1 - 2 347
          Gene Symbol: Igh-V7183
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA D29667.1 - 3 299
          L21016.1 - 3 365
          L21018.1 - 3 365
          L31514.1 - 2 350
          M21520.1 - 3 338
          U62046.1 - 2 314
          Gene Symbol: Igh-VJ558
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK142609.1 - 3 430
          BC013488.1 - 4 420 4215488
          BC013490.1 - 5 423 4216550
          BC019425.1 - 3 467 4459683
          U36491.1 - 317
          U38851.1 - 326
          U38853.1 - 323
          U38854.1 - 311
          U39780.1 - 329
          Gene Symbol: Igh-VS107
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA X14096.1 - 425
          X63806.1 - 2 341
          Gene Symbol: Ighg
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA X70423.1 - 5 467
          Gene Symbol: Ighg3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AY571287.1 - 420
          Gene Symbol: Ighm
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AB067781.1 - 353
          AB067792.1 - 353
          AB067793.1 - 350
          AF052834.1 - 350
          AF052835.1 - 350
          AF321953.1 - 341
          AY172900.1 - 314
          AY182567.1 - 317
          M27756.1 - 134
          M77137.1 - 314
          Z22061.1 - 341
          Gene Symbol: Ighv1-53
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AB069917.1 - 335
          Gene Symbol: Ighv1-54
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AJ006946.1 - 401
          Gene Symbol: Ighv1-61
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AB160430.1 - 353
          Gene Symbol: Ighv1-62
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AY172893.1 - 311
          AY172894.1 - 311
          DQ416709.1 - 321
          HQ446589.1 - 313
          Gene Symbol: Ighv1-72
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AF045489.1 - 398
          J00529.1 - 407
          J00539.1 - 401
          M27788.1 - 443
          M27789.1 - 380
          U88688.1 - 401
          Gene Symbol: Ighv6-6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AB050074.1 - 423
          Gene Symbol: Ighv7-3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AF045506.1 - 401
          EF492998.1 - 350
          JF690437.1 - 301
          JX120342.1 - 344
          Z72464.1 - 353
          Gene Symbol: Igk-V1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA Z95477.1 - 731
          Gene Symbol: LOC102642252
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AY172895.1 - 302
          AY594681.1 - 326
          EU915625.1 - 264
          FJ816492.1 - 275
          FJ816546.1 - 272
          FJ816558.1 - 251
          FJ816595.1 - 272
          FJ816599.1 - 266
          GU126674.1 - 347
          HF569536.1 - 321
          HF569540.1 - 327
          JX492330.1 - 335
          JX492360.1 - 341
          JX657377.1 - 318
          JX657516.1 - 306
          JX657517.1 - 306
          KF208092.1 - 305
          KF208218.1 - 332
          KJ398474.1 - 334
          M28251.1 - 395
          Gene Symbol: LOC105243590
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA FJ816503.1 - 269
          Gene Symbol: LOC238440
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA M23051.1 - 320
          M31908.1 - 353
          M32036.1 - 353
          M32038.1 - 353
          Gene Symbol: LOC382693
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AB069916.1 - 2 344
          Gene Symbol: Rac1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA U25107.1 - 338
          Gene Symbol: Tnpo3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA M18568.1 - 200
          Gene Symbol: V165-D-J-C mu
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AB069915.1 - 2, 3 344
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA U22992.1 - 350 Chr. - - [-] on Build

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          immunoglobulin

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0