Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP58-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108036.1 | 2 | 672 | |||
GenBank mRNA | BG204753.1 | - | 1 | 215 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000435434.1 | - | 2 | 672 | Chr. 10 - 104351591 - 104353575 [-] on Build GRCh38 |