Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CECR5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024483.1 | 1 | 343 | |||
GenBank mRNA | AF307448.1 | - | 1 | 338 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000431923.1 | - | 1 | 333 | Chr. 22 - 17159399 - 17165445 [+] on Build GRCh38 |