siRNA Details
Gene Symbol: LINC00661 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026828.1 | 8 | 912 | |||
GenBank mRNA | BC117476.1 | - | 4 | 536 | ||
BC144048.1 | - | 4 | 514 | |||
CR749400.1 | - | 8 | 912 |

Gene Symbol: LINC00661 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026828.1 | 8 | 912 | |||
GenBank mRNA | BC117476.1 | - | 4 | 536 | ||
BC144048.1 | - | 4 | 514 | |||
CR749400.1 | - | 8 | 912 |