Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC150935 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK056439.1 | - | 4 | 3068 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000407524.1 | - | 3 | 3093 | Chr. 2 - 239762860 - 239802883 [+] on Build GRCh38 |