Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TP73-AS1 SIRNA Assays ›
          Gene Symbol:
          TP73-AS1
          Gene Name:
          TP73 antisense RNA 1
          Gene Aliases:
          KIAA0495, PDAM
          Chromosome Location:
          Chr.1: 3735984 - 3747373 on Build GRCh38
          UniGene ID:
          Hs.49658
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n505462

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TP73-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_033708.1 5 2611
          NR_033709.1 5 3224
          NR_033710.1 5 4479
          NR_033711.1 4 4943
          NR_033712.1 6 1858
          GenBank mRNA AB007964.1 - 4 4893
          AL133648.1 - 5 3732
          BC035073.1 - 5 2576
          BC044603.1 - 4 756
          BC089039.1 - 5 3224
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA CN302812.1 - 1 273 Chr. 1 - 3737028 - 3737712 [-] on Build GRCh38
          GenBankmRNA BX119697.1 - 1 288 Chr. 1 - 3737149 - 3737738 [-] on Build GRCh38
          GenBankmRNA D45464.1 - 1 189 Chr. 1 - 3737248 - 3737639 [-] on Build GRCh38
          GenBankmRNA CB215637.1 - 1 38 Chr. 1 - 3736847 - 3737487 [-] on Build GRCh38
          GenBankmRNA BM725718.1 - 1 219 Chr. 1 - 3737149 - 3737668 [-] on Build GRCh38
          GenBankmRNA CD359309.1 - 1 543 Chr. 1 - 3737170 - 3737992 [-] on Build GRCh38
          GenBankmRNA BQ670338.1 - 1 727 Chr. 1 - 3737212 - 3738162 [-] on Build GRCh38
          GenBankmRNA CA430359.1 - 1 426 Chr. 1 - 3737149 - 3737875 [-] on Build GRCh38
          GenBankmRNA DB519579.1 - 1 156 Chr. 1 - 3737150 - 3737605 [-] on Build GRCh38
          GenBankmRNA DB527897.1 - 1 176 Chr. 1 - 3737150 - 3737625 [-] on Build GRCh38
          GenBankmRNA W69964.1 - 245 Chr. - - [-] on Build
          GenBankmRNA DB545331.1 - 1 157 Chr. 1 - 3737162 - 3737605 [-] on Build GRCh38
          GenBankmRNA BF977691.1 - 567 Chr. - - [-] on Build
          GenBankmRNA BQ429058.1 - 1 526 Chr. 1 - 3737170 - 3737974 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0