Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR4290HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038882.1 | 4 | 2592 | |||
GenBank mRNA | BC039331.1 | - | 4 | 2591 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000426593.1 | - | 4 | 2592 | Chr. 9 - 90020654 - 90041499 [-] on Build GRCh38 |