Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01209 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110819.1 | 1, 2 | 820 | |||
GenBank mRNA | BC041880.1 | - | 1, 2 | 821 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000451289.1 | - | 1, 2 | 820 | Chr. 3 - 176814155 - 176817001 [-] on Build GRCh38 |