Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BOK-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033346.1 | 2 | 757 | |||
GenBank mRNA | FJ185168.1 | - | 2 | 757 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000434306.1 | - | 2 | 591 | Chr. 2 - 241544403 - 241558977 [-] on Build GRCh38 |