Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01103 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110113.1 | 6 | 1140 | |||
GenBank mRNA | BC040913.1 | - | 6 | 1140 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000430551.2 | - | 6 | 1140 | Chr. 2 - 104488458 - 104510509 [-] on Build GRCh38 |