Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C22orf34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017028796.1 | XP_016884285.1 | 7 | 10392 | ||
GenBank mRNA | AK128192.1 | - | 2 | 683 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000420902.1 | - | 2 | 683 | Chr. 22 - 49612657 - 49615716 [-] on Build GRCh38 |