siRNA Details
Gene Symbol: ADAMTS9-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110150.1 | 2 | 610 | |||
GenBank mRNA | DA222486.1 | - | 2 | 289 |

Gene Symbol: ADAMTS9-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110150.1 | 2 | 610 | |||
GenBank mRNA | DA222486.1 | - | 2 | 289 |