siRNA Details
Gene Symbol: ADAMTS9-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110150.1 | 3 | 907 | |||
NR_110151.1 | 2 | 668 | ||||
GenBank mRNA | AK125532.1 | - | 2 | 303 | ||
DA222486.1 | - | 3 | 586 |

Gene Symbol: ADAMTS9-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110150.1 | 3 | 907 | |||
NR_110151.1 | 2 | 668 | ||||
GenBank mRNA | AK125532.1 | - | 2 | 303 | ||
DA222486.1 | - | 3 | 586 |