Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXTL3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC043205.2 | - | 2 | 1551 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000523789.2 | - | 3 | 418 | Chr. 8 - 28698212 - 28700530 [-] on Build GRCh38 |