Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLJ20021 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033874.1 | 2 | 739 | |||
GenBank mRNA | AK000028.1 | - | 2 | 735 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000527564.1 | - | 2 | 736 | Chr. 4 - 101347780 - 101348883 [+] on Build GRCh38 |