Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK4-TEX40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_133662.1 | 7 | 1902 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000539086.5 | - | 7 | 1902 | Chr. 11 - 64291722 - 64304769 [+] on Build GRCh38 |