Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC-ROR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_048536.1 | 4 | 2448 | |||
GenBank mRNA | HQ315778.1 | - | 4 | 2448 | ||
LC027886.1 | - | 5 | 2063 | |||
LC027887.1 | - | 5 | 1262 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000553704.2 | - | 4 | 2448 | Chr. 18 - 57054582 - 57072119 [-] on Build GRCh38 |