Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXN3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036500.1 | 2 | 746 | |||
GenBank mRNA | BC067103.1 | - | 2 | 746 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000555562.1 | - | 2 | 746 | Chr. 14 - 89417354 - 89419793 [+] on Build GRCh38 |