Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AA06 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037584.2 | 3 | 417 | |||
GenBank mRNA | AF381171.1 | - | 3 | 383 | ||
AF381172.1 | - | 3 | 386 | |||
AF381173.1 | - | 3 | 393 | |||
AF381174.1 | - | 3 | 393 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000579745.1 | - | 3 | 417 | Chr. 17 - 33529787 - 33533760 [-] on Build GRCh38 |