Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC728485 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294306.1 | NP_001281235.1 | 2 | 1531 | ||
GenBank mRNA | AK092138.1 | - | 2 | 1510 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000590750.1 | - | 2 | 1496 | Chr. 19 - 36773153 - 36777078 [+] on Build GRCh38 |