Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ERVK-19 SIRNA Assays ›
          Gene Symbol:
          ERVK-19 ERVK-25 ERVK-4 ERVK-6 ERVK-9 LOC105369972 LOC105376906 LOC107986724
          Gene Name:
          endogenous retrovirus group K member 19
          endogenous retrovirus group K member 25
          endogenous retrovirus group K member 4
          endogenous retrovirus group K member 6
          endogenous retrovirus group K member 9
          endogenous retrovirus group K member 8 Pol protein-like
          endogenous retrovirus group K member 19 Pol protein
          endogenous retrovirus group K member 6 Pol protein-like
          Gene Aliases:
          HERV-K(C19), P1.8, PR, Protease, Proteinase, envK3, polymerase
          PR, Protease, Proteinase, c11_A
          ERVK4, HERV-K(I), HERV-KI
          ERVK6, HERV-K(C7), HERV-K108, K-Rev, c-orf, cORF
          PR, Protease, Proteinase, polymerase
          -
          -
          -
          Chromosome Location:
          -
          Chromosome Location:
          -
          Chromosome Location:
          -
          Chromosome Location:
          -
          Chromosome Location:
          -
          Chromosome Location:
          Chr.12: 58323535 - 58331743 on Build GRCh38
          Chromosome Location:
          Chr.19: 27520104 - 27641535 on Build GRCh38
          Chromosome Location:
          Chr.7: 4578493 - 4595492 on Build GRCh38
          UniGene ID:
          -
          -
          -
          -
          -
          -
          -
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID n515031

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ERVK-19
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AY395519.1 - 1 157
          Gene Symbol: ERVK-25
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA CF227268.1 - 2 370
          Gene Symbol: ERVK-4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AY371031.1 - 1 157
          AY371037.1 - 1 157
          AY395520.1 - 1 107
          AY395523.1 - 1 157
          AY395524.1 - 1 157
          AY395525.1 - 1 157
          Gene Symbol: ERVK-6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AY371029.1 - 1 157
          AY371033.1 - 1 157
          AY371034.1 - 1 157
          AY371035.1 - 1 157
          AY371036.1 - 1 157
          AY371042.1 - 1 157
          AY371043.1 - 1 157
          AY371044.1 - 1 157
          AY371045.1 - 1 157
          AY395517.1 - 1 157
          AY395522.1 - 1 157
          Gene Symbol: ERVK-9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AY371030.1 - 1 157
          Gene Symbol: LOC105369972
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_011539093.2 XP_011537395.1 1 1420
          Gene Symbol: LOC105376906
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_017027549.1 XP_016883038.1 1 1054
          Gene Symbol: LOC107986724
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_017012900.1 XP_016868389.1 1 1698
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GENCODE ENST00000600419.5 - 1 157 Chr. 19 - 27638483 - 27640638 [-] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          viral or transposable element protein

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-h2dpj:80/100.66.76.115:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline