Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209390.1 | - | 3 | 5463 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000575331.1 | - | 3 | 5463 | Chr. 17 - 7439506 - 7445966 [+] on Build GRCh38 |