Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trerf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK042320.1 | - | 2 | 382 | ||
AK154542.1 | - | 3 | 253 | |||
AK155049.1 | - | 3 | 355 | |||
AK155404.1 | - | 3 | 254 | |||
AK170333.1 | - | 2 | 243 | |||
AK170342.1 | - | 3 | 349 | |||
BC059215.1 | - | 4 | 241 | 6412734 | ||
BU705739.1 | - | 4 | 241 |