Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trerf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK035176.1 | - | 1 | 226 | ||
AK042320.1 | - | 4 | 760 | |||
AK046822.1 | - | 1 | 97 | |||
AK154542.1 | - | 5 | 629 | |||
AK155049.1 | - | 5 | 734 | |||
AK155404.1 | - | 5 | 630 | |||
AK170342.1 | - | 5 | 725 | |||
AY081774.1 | - | 2 | 369 | |||
BC059215.1 | - | 6 | 619 | 6412734 | ||
BC108416.1 | - | 1 | 79 | 5697521 | ||
BU705739.1 | - | 6 | 619 |