Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ERBB2 SIRNA Assays ›
Gene Symbol:
Erbb2
Gene Name:
v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog (avian)
Gene Aliases:
Erbb-2, HER-2, HER2, Neu, c-erbB2, c-neu, mKIAA3023
Chromosome Location:
Chr.11: 98412484 - 98437716 on Build GRCm38
UniGene ID:
Mm.290822
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
n517210
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
20 nmol
20 nmol
40 nmol
250 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
InVivo | 1 umol
1 nmol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
20 nmol
20 nmol
40 nmol
250 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
InVivo | 1 umol
1 nmol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
20 nmol
20 nmol
40 nmol
250 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
InVivo | 1 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Erbb2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
AK160975.1
-
1680
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
Transcript Location
GenBankmRNA
BI155688.1
-
1
40
Chr. 11 - 98438576 - 98439342 [+] on Build GRCm38
GenBankmRNA
BI156098.1
-
1
413
Chr. 11 - 98438191 - 98439039 [+] on Build GRCm38
GenBankmRNA
BI651638.1
-
103
Chr. - - [-] on Build
GenBankmRNA
AK027956.1
-
31
Chr. - - [-] on Build
GenBankmRNA
BI645894.1
-
1
23
Chr. 11 - 98438580 - 98439476 [+] on Build GRCm38
GenBankmRNA
BI652995.1
-
1
13
Chr. 11 - 98438590 - 98439237 [+] on Build GRCm38
GenBankmRNA
BB609667.1
-
1
31
Chr. 11 - 98438572 - 98439247 [+] on Build GRCm38
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-bp9xj:80/100.66.77.12:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0