Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt61b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015549.1 | 1 | 649 | |||
NR_027952.1 | 1 | 649 | ||||
GenBank mRNA | AK004173.1 | - | 1 | 141 | ||
AK032413.1 | - | 1 | 634 | |||
AK036113.1 | - | 1 | 643 | |||
AK045842.1 | - | 1 | 595 |